accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOBOH

CAS RN 5122-94-1

IUPAC Name: {[1,1'-biphenyl]-4-yl}boronic acid
Synonyms: [1,1'-biphenyl]-4-ylboronic acid
Molecular Weight (g/mol): 198.03
Molecular Formula: C12H11BO2
InChi Key: XPEIJWZLPWNNOK-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results