accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5122-99-6

IUPAC Name: (4-iodophenyl)boronic acid
Synonyms: 4-iodophenylboronic acid
Molecular Weight (g/mol): 247.83
Molecular Formula: C6H6BIO2
InChi Key: PELJYVULHLKXFF-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(I)C=C1
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results