accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NOHH2N(E/Z)

CAS RN 51285-13-3

IUPAC Name: N'-hydroxycyclopropanecarboximidamide
Synonyms: N'-hydroxycyclopropanecarboximidamide
Molecular Weight (g/mol): 100.12
Molecular Formula: C4H8N2O
InChi Key: OMCUPXRCMTUDHI-UHFFFAOYSA-N
SMILES: NC(=NO)C1CC1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (5)
  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)