accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 513-49-5

H3CCH3H2N

CAS RN 513-49-5

IUPAC Name: butan-2-amine
Synonyms: sec-butylamine
Molecular Weight (g/mol): 73.14
Molecular Formula: C4H11N
InChi Key: BHRZNVHARXXAHW-UHFFFAOYNA-N
SMILES: CCC(C)N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results