accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CdOOO

CAS RN 513-78-0

IUPAC Name: cadmium(2+) carbonate
Synonyms: cadmium carbonate
Molecular Weight (g/mol): 172.42
Molecular Formula: CCdO3
InChi Key: GKDXQAKPHKQZSC-UHFFFAOYSA-L
SMILES: [Cd++].[O-]C([O-])=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (3)
  • (2)