accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 51312-42-6

IUPAC Name: sodium tungsten hydroxide oxidandiide phosphate
Synonyms: sodium tungsten hydroxide oxidandiide phosphate
Molecular Weight (g/mol): 334.81
Molecular Formula: HNaO6PW
InChi Key: REGJTQWYOKLGSR-UHFFFAOYSA-J
SMILES: [OH-].[O--].[Na+].[W].[O-]P([O-])([O-])=O
Grade 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)