accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 51317-66-9

IUPAC Name: 4-(4-isothiocyanatophenyl)morpholine
Synonyms: 4-(4-isothiocyanatophenyl)morpholine
Molecular Weight (g/mol): 220.29
Molecular Formula: C11H12N2OS
InChi Key: AXUXRZZYZBZQAR-UHFFFAOYSA-N
SMILES: S=C=NC1=CC=C(C=C1)N1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results