accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NOOOH

CAS RN 51362-30-2

IUPAC Name: 3-(pyridin-2-yloxy)benzoic acid
Synonyms: 3-(pyridin-2-yloxy)benzoic acid
Molecular Weight (g/mol): 215.21
Molecular Formula: C12H9NO3
InChi Key: LYSIEAIIZBZRCE-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(OC2=CC=CC=N2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results