accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 51363-00-9

IUPAC Name: 4-(pyridin-2-yloxy)benzoic acid
Synonyms: 4-(pyridin-2-yloxy)benzoic acid
Molecular Weight (g/mol): 215.21
Molecular Formula: C12H9NO3
InChi Key: GKSKQZLHPWBLJL-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(OC2=CC=CC=N2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results