accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3OOCH3CH3Br

CAS RN 51368-55-9

IUPAC Name: propan-2-yl 2-bromo-2-methylpropanoate
Synonyms: isopropyl 2-bromo-2-methylpropanoate
Molecular Weight (g/mol): 209.08
Molecular Formula: C7H13BrO2
InChi Key: UNZJYKKJZGIFCG-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)C(C)(C)Br
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)