accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5137-70-2

IUPAC Name: dodecylphosphonic acid
Synonyms: phosphonic acid, dodecyl-
Molecular Weight (g/mol): 250.32
Molecular Formula: C12H27O3P
InChi Key: SVMUEEINWGBIPD-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCP(O)(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results