CAS RN 51373-66-1
CAS RN 51373-66-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Terbium(III) oxalate decahydrate, REacton™, 99.9% (REO), Thermo Scientific™
CAS: 51373-66-1 Molecular Formula: C6O12Tb2 Molecular Weight (g/mol): 581.905 MDL Number: MFCD00748140 InChI Key: JZNKXIAEFQDQDF-UHFFFAOYSA-H Synonym: terbium iii oxalate,diterbium trioxalate,edioic acid, terbium 3 salt 3:2,oxalate; terbium 3+,ethanedioic acid, terbium 3+ salt 3:2,acmc-1cs38,terbium, mu-ethanedioato 2--kappao1,kappao2':kappao1',kappao2 bis ethanedioato 2--kappao1,kappao2 di PubChem CID: 164770 IUPAC Name: oxalate;terbium(3+) SMILES: C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].[Tb+3].[Tb+3]