accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 51388-20-6

IUPAC Name: hydrogen 4-(benzyloxy)aniline chloride
Synonyms: hydrogen aniline, p-benzyloxy- chloride
Molecular Weight (g/mol): 235.71
Molecular Formula: C13H14ClNO
InChi Key: KQBDLOVXZHOAJI-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=CC=C(OCC2=CC=CC=C2)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (3)
Quantity 
  • (3)
  • (3)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)