CAS RN 51404-69-4
IUPAC Name:
(acetyloxy)plumbyl acetate dihydrate lead
Synonyms:
lead acetate (II), trihydrate dihydrate lead
Molecular Weight (g/mol):
568.5
Molecular Formula:
C4H10O6Pb2
InChi Key:
XAOPZCQFBUCRKR-UHFFFAOYSA-L
SMILES:
O.O.[Pb].CC(=O)O[Pb]OC(C)=O
1
–
4
of
4
results
1
Lead(II) acetate basic, ACS reagent
CAS: 51404-69-4 Molecular Formula: C4H10O8Pb3 Molecular Weight (g/mol): 807.69 InChI Key: XAOPZCQFBUCRKR-UHFFFAOYSA-L PubChem CID: 75087083 IUPAC Name: diacetyloxylead;lead;dihydrate SMILES: CC(=O)O[Pb]OC(=O)C.O.O.[Pb]
2
Lead Acetate Basic, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.