accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 515-96-8

IUPAC Name: 1-(hydrazinecarbonyl)formamide
Synonyms: 1-(hydrazinecarbonyl)formamide
Molecular Weight (g/mol): 103.08
Molecular Formula: C2H5N3O2
InChi Key: MOKRDWKSHLLYKM-UHFFFAOYSA-N
SMILES: NNC(=O)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results