accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHOOCH3

CAS RN 5150-42-5

IUPAC Name: 2,3-dimethoxyphenol
Synonyms: 2,3-dimethoxyphenol
Molecular Weight (g/mol): 154.17
Molecular Formula: C8H10O3
InChi Key: QSZCGGBDNYTQHH-UHFFFAOYSA-N
SMILES: COC1=CC=CC(O)=C1OC
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)