accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 515158-85-7

IUPAC Name: magnesium(2+) 3-chloro-4-methylbenzen-1-ide bromide
Synonyms: magnesium(2+) 3-chloro-4-methylbenzen-1-ide bromide
Molecular Weight (g/mol): 229.78
Molecular Formula: C7H6BrClMg
InChi Key: XQLVSPRXTWDGNL-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].CC1=CC=[C-]C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results