accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 51517-88-5

ONHNHNN

CAS RN 51517-88-5

IUPAC Name: 4-(2,5-dihydro-1H-1,2,3,4-tetrazol-5-ylidene)cyclohexa-2,5-dien-1-one
Synonyms: 4-(1,2-dihydro-1,2,3,4-tetrazol-5-ylidene)cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 162.15
Molecular Formula: C7H6N4O
InChi Key: GDDQLGTXWIFSDI-UHFFFAOYSA-N
SMILES: O=C1C=CC(C=C1)=C1NNN=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results