accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NNHFF

CAS RN 51523-79-6

IUPAC Name: (2,4-difluorophenyl)hydrazine
Synonyms: (2,4-difluorophenyl)hydrazine
Molecular Weight (g/mol): 144.13
Molecular Formula: C6H6F2N2
InChi Key: RLZUIPTYDYCNQI-UHFFFAOYSA-N
SMILES: NNC1=CC=C(F)C=C1F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results