accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CBr

CAS RN 51554-93-9

IUPAC Name: 1-bromo-4-octylbenzene
Synonyms: 1-bromo-4-octylbenzene
Molecular Weight (g/mol): 269.23
Molecular Formula: C14H21Br
InChi Key: OOZQSVXPBCINJF-UHFFFAOYSA-N
SMILES: CCCCCCCCC1=CC=C(Br)C=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)