accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CBr

CAS RN 51554-95-1

IUPAC Name: 1-bromo-4-pentylbenzene
Synonyms: 1-bromo-4-pentylbenzene
Molecular Weight (g/mol): 227.15
Molecular Formula: C11H15Br
InChi Key: SGCJPYYTVBHQGE-UHFFFAOYSA-N
SMILES: CCCCCC1=CC=C(Br)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (3)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results