accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COONH2

CAS RN 5159-59-1

IUPAC Name: methyl 6-aminonaphthalene-2-carboxylate
Synonyms: methyl 6-aminonaphthalene-2-carboxylate
Molecular Weight (g/mol): 201.23
Molecular Formula: C12H11NO2
InChi Key: LPXVPYIHRFOTJZ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C2C=C(N)C=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)