accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 516-02-9

BaOOOO

CAS RN 516-02-9

IUPAC Name: barium(2+) oxalate
Synonyms: barium oxalate
Molecular Weight (g/mol): 225.35
Molecular Formula: C2BaO4
InChi Key: GXUARMXARIJAFV-UHFFFAOYSA-L
SMILES: [Ba++].[O-]C(=O)C([O-])=O
Grade 
  • (3)
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results