accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrH2C

CAS RN 5162-44-7

IUPAC Name: 4-bromobut-1-ene
Synonyms: 4-bromo-1-butene
Molecular Weight (g/mol): 135
Molecular Formula: C4H7Br
InChi Key: DMAYBPBPEUFIHJ-UHFFFAOYSA-N
SMILES: BrCCC=C
Molecular Weight (g/mol) 
  • (6)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)