accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 51626-33-6

IUPAC Name: 5-chloro-3-methyl-1-phenyl-1H-pyrazole-4-carbonitrile
Synonyms: 5-chloro-3-methyl-1-phenylpyrazole-4-carbonitrile
Molecular Weight (g/mol): 217.66
Molecular Formula: C11H8ClN3
InChi Key: MWXQWRGAGVHIDR-UHFFFAOYSA-N
SMILES: CC1=NN(C(Cl)=C1C#N)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results