accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 51642-03-6

IUPAC Name: hydrogen 3-amino-1λ⁶-thiolane-1,1-dione chloride
Synonyms: hydrogen 3-amino-1λ⁶-thiolane-1,1-dione chloride
Molecular Weight (g/mol): 171.64
Molecular Formula: C4H10ClNO2S
InChi Key: MGZQMSFXPSKBDY-UHFFFAOYNA-N
SMILES: [H+].[Cl-].NC1CCS(=O)(=O)C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)