accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 51673-83-7

IUPAC Name: oxane-2-carboxylic acid
Synonyms: oxane-2-carboxylic acid
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: MQAYFGXOFCEZRW-UHFFFAOYNA-N
SMILES: OC(=O)C1CCCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results