accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 517-22-6

IUPAC Name: 3-ethyl-2,4-dimethyl-1H-pyrrole
Synonyms: 3-ethyl-2,4-dimethyl-1H-pyrrole
Molecular Weight (g/mol): 123.2
Molecular Formula: C8H13N
InChi Key: ZEBBLOXDLGIMEG-UHFFFAOYSA-N
SMILES: CCC1=C(C)NC=C1C
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results