accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

(S)(R)OOHOHHOOHHOH

CAS RN 517-28-2

IUPAC Name: (1S,10R)-8-oxatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-2,4,6,12(17),13,15-hexaene-5,6,10,14,15-pentol
Synonyms: hematoxylin (-)-haematoxylin
Molecular Weight (g/mol): 302.28
Molecular Formula: C16H14O6
InChi Key: WZUVPPKBWHMQCE-CJNGLKHVSA-N
SMILES: OC1=CC=C2[C@@H]3C4=C(C[C@]3(O)COC2=C1O)C=C(O)C(O)=C4
Quantity 
  • (8)
  • (1)
  • (8)
  • (1)
  • (4)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (17)
Color 
  • (3)
  • (2)
  • (6)
  • (1)
Percent Purity 
  • (3)
  • (3)
  • (3)
Grade 
  • (2)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (3)
  • (6)
  • (6)
  • (1)
  • (3)
  • (8)
  • (1)
  • (8)
  • (1)
  • (4)
  • (2)
  • (1)
  • (3)
  • (17)
  • (3)
  • (2)
  • (6)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (6)
  • (4)