accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 51707-55-2

IUPAC Name: 1-phenyl-3-(1,2,3-thiadiazol-5-yl)urea
Synonyms: thidiazuron
Molecular Weight (g/mol): 220.25
Molecular Formula: C9H8N4OS
InChi Key: HFCYZXMHUIHAQI-UHFFFAOYSA-N
SMILES: O=C(NC1=CN=NS1)NC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results