accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 51721-39-2

H3COH

CAS RN 51721-39-2

IUPAC Name: dec-3-yn-1-ol
Synonyms: dec-3-yn-1-ol
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: YGEQBZUDPQQIFI-UHFFFAOYSA-N
SMILES: CCCCCCC#CCCO
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)