accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 51776-71-7

FFBr

CAS RN 51776-71-7

IUPAC Name: 1-bromo-4-(difluoromethyl)benzene
Synonyms: 1-bromo-4-(difluoromethyl)benzene
Molecular Weight (g/mol): 207.02
Molecular Formula: C7H5BrF2
InChi Key: HUSPSWKWFREKSS-UHFFFAOYSA-N
SMILES: FC(F)C1=CC=C(Br)C=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)