accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 518-17-2

IUPAC Name: (1S)-21-methyl-3,13,21-triazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-2(10),4,6,8,15,17,19-heptaen-14-one
Synonyms: evodiamine d-evodiamine (+)-evodiamine evodiamine, (+)-
Molecular Weight (g/mol): 303.37
Molecular Formula: C19H17N3O
InChi Key: TXDUTHBFYKGSAH-SFHVURJKSA-N
SMILES: CN1[C@H]2N(CCC3=C2NC2=CC=CC=C32)C(=O)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Product Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results