accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 518-47-8

NaNaOOOOO(?)

CAS RN 518-47-8

IUPAC Name: disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Synonyms: disodium 2-(3-oxido-6-oxoxanthen-9-yl)benzoate
Molecular Weight (g/mol): 376.28
Molecular Formula: C20H10Na2O5
InChi Key: NJDNXYGOVLYJHP-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Color 
  • (3)
  • (3)
Grade 
  • (3)
  • (3)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)