accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 518-75-2

IUPAC Name: (3R,4S)-7-(dihydroxymethylidene)-3,4,5-trimethyl-4,6,7,8-tetrahydro-3H-2-benzopyran-6,8-dione
Synonyms: citrinin
Molecular Weight (g/mol): 250.25
Molecular Formula: C13H14O5
InChi Key: JLWQFKMKUKQXMW-IYSWYEEDSA-N
SMILES: C[C@H]1OC=C2C(=O)C(=C(O)O)C(=O)C(C)=C2[C@@H]1C
Quantity 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)