accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3FOHO

CAS RN 518070-22-9

IUPAC Name: 2-(3-fluoro-5-methylphenyl)acetic acid
Synonyms: (3-fluoro-5-methylphenyl)acetic acid
Molecular Weight (g/mol): 168.17
Molecular Formula: C9H9FO2
InChi Key: GDUUQFGFHKTVEB-UHFFFAOYSA-N
SMILES: CC1=CC(CC(O)=O)=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results