accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 51850-20-5

IUPAC Name: dihydrogen platinumhexakis(olate)
Synonyms: dihydrogen platinumhexakis(olate)
Molecular Weight (g/mol): 299.14
Molecular Formula: H8O6Pt
InChi Key: BYFKUSIUMUEWCM-UHFFFAOYSA-J
SMILES: [H+].[H+].[OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Pt]
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results