accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5187-82-6

IUPAC Name: (2-ethoxy-2-oxoethyl)dimethylsulfanium bromide
Synonyms: (2-ethoxy-2-oxoethyl)dimethylsulfanium bromide
Molecular Weight (g/mol): 229.13
Molecular Formula: C6H13BrO2S
InChi Key: JXFPTJYKYKVENJ-UHFFFAOYSA-M
SMILES: [Br-].CCOC(=O)C[S+](C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results