CAS RN 519-02-8
CAS RN 519-02-8
IUPAC Name:
(1R,2R,9S,17S)-7,13-diazatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadecan-6-one
Synonyms:
matrine matrene, (+)- (5β)-isomer matrine sophoridine matrine, (5β)-isomer (+)-matrine (+)-matrene (5β)-matrine matridin-15-one (9CI) matrine, (5β)-
Molecular Weight (g/mol):
248.37
Molecular Formula:
C15H24N2O
InChi Key:
ZSBXGIUJOOQZMP-JLNYLFASSA-N
SMILES:
O=C1CCC[C@@H]2[C@H]3CCCN4CCC[C@@H](CN12)[C@@H]34
1
–
2
of
2
results
1
Thermo Scientific Chemicals Matrine
CAS: 519-02-8 Molecular Formula: C15H24N2O
2
Matrine 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality