accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 519-73-3

IUPAC Name: (diphenylmethyl)benzene
Synonyms: triphenylmethane
Molecular Weight (g/mol): 244.34
Molecular Formula: C19H16
InChi Key: AAAQKTZKLRYKHR-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (6)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)