accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 519056-67-8

IUPAC Name: 1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-sulfonyl chloride
Synonyms: 1-methyl-3-(trifluoromethyl)pyrazole-4-sulfonyl chloride
Molecular Weight (g/mol): 248.6
Molecular Formula: C5H4ClF3N2O2S
InChi Key: HJFWSNIHZZKYIK-UHFFFAOYSA-N
SMILES: CN1C=C(C(=N1)C(F)(F)F)S(Cl)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results