accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CPOOCH3OCH3

CAS RN 5191-09-3

IUPAC Name: diethyl tetradecylphosphonate
Synonyms: diethyl tetradecylphosphonate
Molecular Weight (g/mol): 334.48
Molecular Formula: C18H39O3P
InChi Key: CPWIYYOBPHXJIA-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCP(=O)(OCC)OCC
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results