accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5198-88-9

IUPAC Name: 2-bromo-1,3-thiazole-4-carboxylate
Synonyms: 2-bromo-1,3-thiazole-4-carboxylate
Molecular Weight (g/mol): 207.02
Molecular Formula: C4HBrNO2S
InChi Key: BEGREHRAUWCAHV-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CSC(Br)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results