accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SPNNN

CAS RN 52-24-4

IUPAC Name: tris(aziridin-1-yl)-λ⁵-phosphanethione
Synonyms: thiophosphamide thiotepa (product) tespa brand of thiotepa thio tepa triethylenethiophosphoramide thiotepa preparation triethylene thiophosphoramide thioplex thio-tepa tespa
Molecular Weight (g/mol): 189.22
Molecular Formula: C6H12N3PS
InChi Key: FOCVUCIESVLUNU-UHFFFAOYSA-N
SMILES: S=P(N1CC1)(N1CC1)N1CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results