accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 52-51-7

IUPAC Name: 2-bromo-2-nitropropane-1,3-diol
Synonyms: bronopol
Molecular Weight (g/mol): 199.99
Molecular Formula: C3H6BrNO4
InChi Key: LVDKZNITIUWNER-UHFFFAOYSA-N
SMILES: OCC(Br)(CO)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (7)
Quantity 
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (4)
  • (7)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)