CAS RN 52-66-4
CAS RN 52-66-4
IUPAC Name:
2-amino-3-methyl-3-sulfanylbutanoic acid
Synonyms:
(+-)-penicillamine penicillamine DL- DMC DL-3-mercaptovaline pencillamine β-mercaptovaline 3,3-dimethylcysteine D,L-penicillamine DL-β-mercaptovaline
Molecular Weight (g/mol):
149.21
Molecular Formula:
C5H11NO2S
InChi Key:
VVNCNSJFMMFHPL-UHFFFAOYNA-N
SMILES:
CC(C)(S)C(N)C(O)=O
1
–
1
of
1
results
1
DL-Penicillamine, 97+%
CAS: 52-66-4 Molecular Formula: C5H11NO2S Molecular Weight (g/mol): 149.21 MDL Number: MFCD00004856 InChI Key: VVNCNSJFMMFHPL-UHFFFAOYNA-N Synonym: dl-penicillamine,2-amino-3-mercapto-3-methylbutanoic acid,+--penicillamine,3,3-dimethyl-dl-cysteine,3-sulfanylvaline,dl-beta-mercaptovaline,beta-mercaptovaline,d,l-penicillamine,dl-3-mercaptovaline,2-amino-3-mercapto-3-methylbutyric acid PubChem CID: 4727 ChEBI: CHEBI:50868 IUPAC Name: 2-amino-3-methyl-3-sulfanylbutanoic acid SMILES: CC(C)(S)C(N)C(O)=O