accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 52-76-6

H3C(S)(S)(R)(R)(S)(R)HOHC

CAS RN 52-76-6

IUPAC Name: (1R,3aS,3bR,9aR,9bS,11aS)-1-ethynyl-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-ol
Synonyms: lynestrenol lynestrenolum lynstranol exlution 17-α-ethynyloestrenol orgametril orgametrol endometril lynoestrenol ethinylestrenol
Molecular Weight (g/mol): 284.44
Molecular Formula: C20H28O
InChi Key: YNVGQYHLRCDXFQ-XGXHKTLJSA-N
SMILES: C[C@]12CC[C@H]3[C@@H](CCC4=CCCC[C@H]34)[C@@H]1CC[C@@]2(O)C#C
Quantity 
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results