accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2SHOHO

CAS RN 52-90-4

IUPAC Name: 2-amino-3-sulfanylpropanoic acid
Synonyms: cysteine zystein Hcys cisteina
Molecular Weight (g/mol): 121.15
Molecular Formula: C3H7NO2S
InChi Key: XUJNEKJLAYXESH-UHFFFAOYNA-N
SMILES: NC(CS)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (16)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (4)
  • (3)
Grade 
  • (1)
  • (4)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (7)
  • (4)
  • (5)
  • (3)
  • (3)
  • (1)
  • (16)
  • (4)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (4)
  • (3)
  • (1)
  • (4)
  • (2)
  • (1)