accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 52009-14-0

IUPAC Name: calcium 2-oxopropanoate
Synonyms: calcium pyruvate
Molecular Weight (g/mol): 127.13
Molecular Formula: C3H3CaO3
InChi Key: QJYMFPMPYYGIJD-UHFFFAOYSA-M
SMILES: [Ca++].CC(=O)C([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)