accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONNNCS

CAS RN 52024-29-0

IUPAC Name: 4-(5-isothiocyanatopyridin-2-yl)morpholine
Synonyms: 4-(5-isothiocyanatopyridin-2-yl)morpholine
Molecular Weight (g/mol): 221.28
Molecular Formula: C10H11N3OS
InChi Key: VLZWMPRZGPULIU-UHFFFAOYSA-N
SMILES: S=C=NC1=CN=C(C=C1)N1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results